Hematology analyser, 3-part differential, BeneSphera™ H-31
Supplier: Avantor
BeneSphera™
BENP20001747EA 245000 INR
BENP20001747
Hematology analyser, 3-part differential, BeneSphera™ H-31
Clinical Diagnostic Supplies Clinical Analysers
BeneSphera™ H-31 is a fully automated, compact and flexible haematology analyser with a throughput of 30 samples per hour and needing only 13 µl of whole blood sample volume to provide 3-part differentiation of WBC, 19 parameters and 3 histograms.
  • Provides result in 2 minutes
  • Storage for up to 10 000 sample results (including histograms)
  • Three counting modes: CBC, WBC/HGB, RBC/PLT
  • Cyanide-free reagents avoid environmental contamination
  • Automatic reagent residual system
  • Built-in thermal recorder (50 mm width paper) and colour LCD display
  • Automatic internal and external sample probe cleaning