2-Phenoxyethanol, J.T.Baker®
Supplier: Avantor
|
Warning
|
Management of Change (MOC) category = N
T319-07EA
29274.68
INR
T319-07
T319-09
2-Phenoxyethanol, J.T.Baker®
2-Phenoxyethanol
Formula:
C₆H₅OCH₂CH₂OH Boiling Pt: 245 °C (1013 hPa) Melting Pt: 13 °C Density: 1,1094 g/cm³ (20 °C) Flash Pt: 121 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002857 CAS Number: 122-99-6 EINECS: 204-589-7 Merck Index: 12,07410 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...