VWR®Signature™, Forced air safety ovens
Supplier: VWR Collection
52201-214 52201-216
VWRU52201-214EA 0 INR
VWRU52201-214 VWRU52201-216
VWR®Signature™, Forced air safety ovens
Ovens Standard Ovens
These microprocessor-controlled forced air safety ovens have heavy gauge steel exteriors and continuous welded seams and corners for long-term durability. The powder coat enamel exterior provides a high resistance to moisture, oil and chemicals.
  • Over-temperature safety limit, manual restart
  • Rear blow-out panel
The panel is designed to release in the event of an explosion, directing the force away from the user. A resettable circuit breaker prevents accidental setting changes in case of electrical malfunction. An over-temperature safety limit requires manual restart to protect both the user and unit.
Other features include electronic timer, heavy-duty adjustable door latch and hinges and 76 mm Ø exhaust port with adjustable damper.
Delivery information: Supplied with three-prong power cord and two stainless steel shelves.
Caution: Unit is not explosion-proof and will not prevent combustibles from exploding. The rear blow-out panel is only an added safety measure. Always use extreme caution when mixing heat and any source of volatile substances in any oven. Use with any volatile substance, material, or residual fumes beyond flashpoint may cause substances to ignite while placed in any oven. Check with your chemical source to ensure safe operational temperature ranges and mixtures while using in conjunction with any heat source. Always exercise extreme caution when handling combustible materials.