VWR®, Deep Well Plates, 12-Well, 22 ml
Supplier: VWR
VWR®
VWRU82007-294EA 0 INR
VWRU82007-294
VWR®, Deep Well Plates, 12-Well, 22 ml
Microplates Deep Well Microplates
These deep-well reservoir plates feature printed alphanumeric labelling and smooth inside surfaces to ensure that all the sample can be pipetted.
  • Compatible with robotic processes
  • Freezable to –90 °C
  • Can be sealed using film or mats
  • Certified free of RNase, DNase, and human DNA