VWR®, pH buffer pouches, NIST, SINGLET™
Supplier: VWR Collection
89236-552
89236-550
89236-548
VWRU89236-552EA
0
INR
VWRU89236-552
VWRU89236-550
VWRU89236-548
VWR®, pH buffer pouches, NIST, SINGLET™
Buffers
pH Calibration Buffers
These pH buffers used for sympHony® pH meter standardisation available in sealed foil pouches.
- Ideal for on-site analyses
- Easy to use
- Fresh and reliable