VWR®, Electrode filling solution
Supplier: Avantor
VWR®
VWRU89236-504EA 0 INR
VWRU89236-504
VWR®, Electrode filling solution
Electrolytes Electrode Filling Solutions
This 3M KCl filling solution for appropriate sympHony® Ag/AgCl reference elements.
  • Compatible with waterproof electrodes
  • Easy to dispense
  • Reliable internal reference