Gram color staining kit microbiology stain
Supplier: VWR Collection
10143-130
10143-606
VWRU10143-130EA
9124
INR
VWRU10143-130
VWRU10143-606
Gram color staining kit microbiology stain
Gram color staining kit
Gram stain sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
Packaging: In styrofoam trays and include spout caps for ease of pouring.