Sand, BDH®
Supplier: BDH (US PRODUCED)
Synonyms:
Silicium dioxide, Silicium (IV) oxide, Silicon dioxide
9274-2.5KG
9274-500G
BDHB9274-2.5KGEA
0
INR
BDHB9274-2.5KG
BDHB9274-500G
Sand, BDH®
Sand
Beige granules.
Synonyms: Quartz, silicon dioxide, silicium dioxide, silicium (Ⅳ) oxide.
Formula:
SiO₂ MW: 60,08 g/mol Boiling Pt: 2230 °C Melting Pt: 1427 °C Density: 2,622…2.645 g/cm³ (20 °C) |
MDL Number:
MFCD02100519 CAS Number: 7631-86-9 |
Specification Test Results
Moisture | ≤0,1% |
Particle size (30 - 40 mesh) | ≥80% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...