J.T.Baker® BAKER INSTRA-ANALYZED®, Sodium Standard Solution, 1000 µg/L Na, reagent for AAS
Supplier: Avantor
BAKR6468-04EA 0 INR
BAKR6468-04
J.T.Baker® BAKER INSTRA-ANALYZED®, Sodium Standard Solution, 1000 µg/L Na, reagent for AAS
Sodium standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
Certifications: Traceable to NIST and SRM numbers.