Nucleic acid modifying kit, Amine, Label IT®
Supplier: MIRUS
MIR3900
MIR3925
MIRUMIR3900EA
0
INR
MIRUMIR3900
MIRUMIR3925
Nucleic acid modifying kit, Amine, Label IT®
Assays
Nucleic Acid Assays
The Label IT® Nucleic Acid Modifying Kits are suitable for direct, non-enzymatic attachment of functional groups to DNA and RNA.
- Direct, covalent attachment of amine functional groups to any nucleic acid
- Easily and consistently control the density of nucleic acid modification
- Permanent, non-destructive modification of nucleic acids, which can then be conjugated to proteins or peptides, or attached to glass surfaces, or beads or plates
Delivery information: Each kit contains Label IT® Reagent, Label IT® Reconstitution Solution, labeling buffer, solutions and G50 spin columns.