Ethyl acetate ≥99.5%

Supplier: VWR Chemicals
Danger

N774-1LEA 7533 INR
N774-1L
Ethyl acetate ≥99.5%
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88,11 g/mol
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0,902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3,II
https://in.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg

Order Now

Specification Test Results

Identification PASS
Purity (GC) = 99,5 %
Specific Gravity @25C 0,894 - 0,898
Water (KF) = 0,2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR