Polyethylene glycol 8,000, crystalline powder, J.T.Baker®
Supplier: Avantor
U222-08EA
46638.68
INR
U222-08
U222-01
U222-09
Polyethylene glycol 8,000, crystalline powder, J.T.Baker®
Polyethylene glycol 8,000
Powder.
Formula:
H(OCH₂CH₂)nOH Boiling Pt: 250 °C (1013 hPa) Density: 1,125 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01779615 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 12,07729 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | Passes Test |
Appearance | Passes Test |
Viscosity, cSt | 470 - 900 |
Average Molecular Weight | 7000 - 9000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...