Sodium dodecyl sulphate (SDS) ≥95%, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
L050-07EA
0
INR
L050-07
L050-02
Sodium dodecyl sulphate (SDS) ≥95%, J.T.Baker®
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na MW: 288,38 g/mol Melting Pt: 204…207 °C Density: 1,1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 2926 ADR: 4.1, 6.1,II Merck Index: 14,08636 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use |
|
Assay (as C₁₂H₂₅OSO₃Na) | ≥95 % |
Identification (by IR) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...