Barium chloride, anhydrous ≥98.0%, powder, purified, J.T.Baker®

Supplier: Avantor
Danger

0980-01EA 15546.77 INR
0980-01
Barium chloride, anhydrous ≥98.0%, powder, purified, J.T.Baker®
Barium chloride

Powder.

Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: ∼963 °C
Density: 3.86 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1 III
Merck Index: 12,00998

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥98.0%
Assay (BaCl₂) ≥98.0 %
Insoluble Matter ≤0.02 %
pH of 5% Solution at 25°C 4.5 - 7.5
Heavy Metals (as Pb) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR