4-(Dimethylamino)benzaldehyde, AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

1836-57EA 0 INR
1836-57 MACR1836-55
4-(Dimethylamino)benzaldehyde, AR® ACS, Macron Fine Chemicals™
4-(Dimethylamino)benzaldehyde
Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (1013 hPa)
Melting Pt: 72
Density: 1.0254 g/cm³ (99.9 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements Passes Test
For Laboratory,Research,or Manufacturing Use Passes Test
ACS - Solubility in Alcohol Passes Test
ACS - Appearance (white to yellowish-white crystals or crystalline powder) Passes Test
ACS - Color of HCl Solution Passes Test
ACS - Melting Point 73 - 75 °C
ACS - Residue after Ignition ≤0.1 %
ACS - Solubility in HCl Passes Test
ACS - Color of Alcohol Solution (APHA) ≤60

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR