n-Pentane ≥98%, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Pentane
T007-09EA
39194.65
INR
T007-09
T007-07
T007-01
n-Pentane ≥98%, BAKER ANALYZED®, J.T.Baker®
n-Pentane
Lot analysis on label.
Formula:
H₃C(CH₂)₃CH₃ MW: 72,15 g/mol Boiling Pt: 35…36 °C (1013 hPa) Melting Pt: –130 °C Density: 0,63 g/cm³ (20 °C) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II REACH: 01-2119459286-30 Merck Index: 13,07193 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water) | ≥98 % |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤0.0020 % |
Sulfur Compounds (as S) | ≤0.005 % |
Water (by KF, coulometric) | ≤0.02 % |
Boiling Range (95%)Lower | 34 - 36 °C |
Boiling Range (95%)Upper | 34 - 40 °C |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...