VWR®, Funnels, Powder
Supplier: VWR Collection
30252-955 30252-953 30252-957
VWRU30252-955EA 0 INR
VWRU30252-955 VWRU30252-953 VWRU30252-957
VWR®, Funnels, Powder
Funnels Powder Funnels
PP, translucent.
  • Parallel stem reduces powder bridging
  • External ribs prevents airlock
  • Resistant to most chemicals except strong oxidizing agents
  • Autoclavable
Used for transferring powders. Used from 0 to 135 °C.