VWR®, Petri Dishes, Disposable
Supplier: VWR Collection
25384-168 25384-318 25384-094 25384-322 25384-308 25384-302 89107-632 25384-306 25384-088 25384-326 25384-092 25384-093 25384-320 25384-328 25384-338 25384-342 25384-344 89022-320 89230-472 25384-090 89137-760 25384-164 25384-310 10799-192 89038-970 89038-968 470210-568 25384-095 25384-312 25384-324 25384-348 10053-704 10799-194 25384-304 25384-346 25384-332 25384-316 25384-166 25384-314 25384-062
VWRU25384-168EA 0 INR
VWRU25384-168 VWRU25384-318 VWRU25384-094 VWRU25384-322 VWRU25384-308 VWRU25384-302 VWRU89107-632 VWRU25384-306 VWRU25384-088 VWRU25384-326 VWRU25384-092 VWRU25384-093 VWRU25384-320 VWRU25384-328 VWRU25384-338 VWRU25384-342 VWRU25384-344 VWRU89022-320 VWRU89230-472 VWRU25384-090 VWRU89137-760 VWRU25384-164 VWRU25384-310 VWRU10799-192 VWRU89038-970 VWRU89038-968 VWRU470210-568 VWRU25384-095 VWRU25384-312 VWRU25384-324 VWRU25384-348 VWRU10053-704 VWRU10799-194 VWRU25384-304 VWRU25384-346 VWRU25384-332 VWRU25384-316 VWRU25384-166 VWRU25384-314 VWRU25384-062
VWR®, Petri Dishes, Disposable
Culture Dishes Petri Dishes
High quality virgin PS, clear for optimum flatness and high optical clarity. Ideal for microbial culture, where accurate bacterial count or microscopic examination is required.
  • Distortion-free visibility
  • Available in varied sizes and styles for specialised and routine applications
  • Stacking ring and slip-ring features available
VWR® Petri dishes are available in a variety of sizes and styles to accommodate both specialised and routine needs. The stacking ring feature, on some models, facilitates positive stacking while the slip-ring feature, on otherMore Product Information