VWR®, Beakers, low form, PMP
Supplier: VWR Collection
89044-514 89044-522 89044-516
VWRU89044-514EA 0 INR
VWRU89044-514 VWRU89044-522 VWRU89044-516
VWR®, Beakers, low form, PMP
Beakers
PMP, crystal clear, graduated.
  • Excellent chemical resistance
  • Autoclavable
  • Withstand a temperature of up to 170 °C for short periods
With large spout and easy to read blue graduations.
Certifications: Conform to ISO 7056 1981 (E) and BS 5404 Part 1.
Order Now
SPECIFICATIONS
Environmentally Preferable