VWR®, Dishes, Crystallising
Supplier: VWR Collection
89000-286 89000-294 89000-280 89000-296
VWRU89000-286EA 0 INR
VWRU89000-286 VWRU89000-294 VWRU89000-280 VWRU89000-296
VWR®, Dishes, Crystallising
Trays/Dishes/Pots Crystallising Dishes
Borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for storage and crystallisation. Rims reinforced and fire-polished to reduce chipping.