VWR®, Measuring Cylinders, Tall Form, Class B, Borosilicate Glass
Supplier: VWR Collection
89000-254 89000-262 89000-260 89000-248 89000-252
VWRU89000-254EA 0 INR
VWRU89000-254 VWRU89000-262 VWRU89000-260 VWRU89000-248 VWRU89000-252
VWR®, Measuring Cylinders, Tall Form, Class B, Borosilicate Glass
Cylinders Measuring Cylinders
Borosilicate glass, tall form, class B.
  • Calibrated to contain at 20 °C ( TC)
  • Graduations printed in durable white enamel
  • Hexagonal base provides stability
Glass conforming to ASTM E1272, Style 1, Class B requirements.
Delivery information: Supplied with a LDPE SAFE-GARD® bumper on sizes 25 ml and larger.