VWR®, Covers and Pans for Test Sieves
Supplier: VWR Collection
57334-662 57334-640 57334-648 57334-656 57334-660 57334-654 57334-658
VWRU57334-662EA 0 INR
VWRU57334-662 510-1579 510-1580 VWRU57334-656 VWRU57334-660 VWRU57334-654 VWRU57334-658
VWR®, Covers and Pans for Test Sieves
Trays/Dishes/Pots Laboratory Trays/Dishes
These durable 8" diameter (203,2 mm) pans and covers fit all VWR® 8" diameter test sieves.
  • Half height (25,4 mm) or full height (50,8 mm) pans are available in brass or stainless steel
  • Pans with extended rims can be used for running multiple tests simultaneously