VWR®, Screw Thread Vials, 9-425
Supplier: VWR Collection
46610-724 46610-722 89524-190
VWRU46610-724EA 0 INR
VWRU46610-724 VWRU46610-722 VWRU89524-190
VWR®, Screw Thread Vials, 9-425
Vials
Borosilicate glass vials ideal for use with Agilent® 7673 and 1100 autosamplers. They are specifically designed with a crimp cap profile for robotic arm autosamplers but have easy-to-use polypropylene screw-thread caps.
Delivery information: Preassembled kits are supplied in one-compartment trays. Unassembled kits are supplied in two-compartment benchtop storage trays.
Order Now
SPECIFICATIONS
Environmentally Preferable