VWR® Labmax, Bottle-Top Dispensers
Supplier: VWR Collection
40000-067 40000-065
VWRU40000-067EA 0 INR
VWRU40000-067 VWRU40000-065
VWR® Labmax, Bottle-Top Dispensers
Bottle Top Dispensers
Variable volume bottle-top dispenser with locking rocker switch, autoclavable.
  • Unique closed circuit air purging system for air and bubbles elimination
  • Exclusive integrated swivel neck
  • Reagent 'guard valve' for safety shut down
  • Patented direct displacement PTFE piston
  • Auto-calibrating, quick and accurate volume adjustment
The dispensers can be disassembled quickly and easily for thorough cleaning. Quick and accurate volume adjustment is easy with the locking rocker switch. The zero point is manufactured into the Labmax™; calibration is not necessary, even after disassembly and cleaning. ISO 9001 Certification has been achieved in the design, development and manufacturing stages of the Labmax™. All Labmax™ dispensers feature a dated mark for compliance with ISO 9001 procedures.