Tank blotters, PerfectBlue™ Web™
Supplier: VWR Peqlab
700-0446EA 0 INR
PEQL52-WEB-10
Tank blotters, PerfectBlue™ Web™
The Web™ S Tank Electroblotter is designed to rapidly transfer nucleic acid or protein fragments from up to four polyacrylamide gels at one time to nitrocellulose, nylon or PVDF membranes. The colour coded cassettes allow for easy assembly of transfer sandwich and error free transfer. The large buffer capacity and integral cooling permit extended transfer times and high voltage/high current operation for the transfer of high molecular weight proteins.
  • Sturdy buffer chamber for up to four colour coded blotting cartridges, with sufficient volume for magnetic stirrer to ensure uniform temperature and pH conditions
  • Highly durable, high-grade steel and platinum-coated titanium electrodes give uniform electrical field over the entire transfer surface; gold-plated plugs and corrosion proof electrodes for long durability and reliable electrical contact
  • Water circulation system for buffer cooling at the base of the tank with effective heat exchanger made of ceramic aluminium and increased exchange surface area to aid heat transfer
  • Safety cover includes securely attached cables to enhance user safety