Trifluoroacetic acid 99, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
TFA
W729-07EA
94217.91
INR
W729-07
W729-05
Trifluoroacetic acid 99, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Trifluoroacetic acid
Lot analysis on label.
Formula:
C₂HF₃O₂ MW: 114.02 g/mol Boiling Pt: 71.8 Melting Pt: –15 °C Density: 1.490 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8 I Merck Index: 13,09754 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use |
|
Assay (F₃CCOOH) | ≥ 99.0 % |
Residue after Evaporation | ≤ 0.0100 % |
Heavy Metals (as Pb) | ≤ 0.001 % |
Trace Impurities - Iron (Fe) | ≤ 5 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...