Sand, dried, washed, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
7062-06EA
0
INR
7062-06
Sand, dried, washed, Macron Fine Chemicals™
Sand
Sand washed and dried.
Caution: For laboratory, research, or manufacturing use.
Formula:
SiO₂ MW: 60,08 g/mol Melting Pt: 1427 °C Storage Temperature: Ambient |
MDL Number:
MFCD02100519 CAS Number: 7631-86-9 EINECS: 238-878-4 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Substances Soluble in HCl | ≤0.16 % |
Appearance (fine, off-white to gray granules) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...