Digital microdispensers
Supplier: DRUMMOND
3-000-525 3-000-550 3-000-575 3-000-590
DRUM3-000-525EA 0 INR
DRUM3-000-525 DRUM3-000-550 DRUM3-000-575 DRUM3-000-590
Digital microdispensers
Pipettes Repetitive Pipettes
Positive displacement microdispensers with direct digital setting and readout for fractional microliters.
  • Positive lock prevents accidental volume changes
  • Overall volumetric accuracy of ±1% (at full volume)
  • Borosilicate glass bores with stainless steel plungers, PP bore on 1000 μl model
  • PTFE tips on all plungers 25 μl or larger