Methanol ACS

Supplier: BDH (US PRODUCED)
Danger

Synonyms: Hydroxy methaneMethyl alcoholCarbinolWood alcohol

BDHB1135-1LPEA 0 INR
BDHB1135-1LP BDHB1135-19L
Methanol ACS
Methanol
Formula: CH₃OH
MW: 32,04 g/mol
Boiling Pt: 64,6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0,7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://in.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR