VWR® Condensers, Coil, Graham Type
Supplier: VWR Collection
201-0588EA 0 INR
201-0588 201-0589 201-0590
VWR® Condensers, Coil, Graham Type
Condensers
Borosilicate glass, grade 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes