VWR® Nessler Cylinders
Supplier: VWR Collection
612-4268EA 1323 INR
612-4268 612-4269
VWR® Nessler Cylinders
Tubes Nessler Tubes
Nessler cylinders for colour comparison.
  • Made of specially selected borosilicate glass 3,3
  • Class A, graduated