VWR® Condensers, Allihn, with Standard Ground Joints
Supplier: VWR Collection
201-0584EA 482 INR
201-0584 201-0585 201-0586 201-0587
VWR® Condensers, Allihn, with Standard Ground Joints
Condensers
Borosilicate glass, grade 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With glass and plastic screw thread connector.
Certifications: ASTM E-438 type 1 Class A.