VWR® Burettes, PTFE Stopcock
Supplier: VWR Collection
612-4262EA 2952 INR
612-4262 612-4263 612-4264
VWR® Burettes, PTFE Stopcock
Burettes
Borosilicate glass 3,3, class B.
Burette with detachable PTFE stopcock, Schellbach stripe.
As per DIN ISO 385 and USP standards.