VWR® Burettes, PTFE Key Stopcock
Supplier: VWR Collection
612-4259EA 1935 INR
612-4259 612-4260 612-4261
VWR® Burettes, PTFE Key Stopcock
Burettes
Borosilicate glass 3,3, class AS.
Clear glass straight bore stopcock with PTFE key, and with LOT certificate.
Accuracy as per class AS DIN 12700, ISO 385.