VWR® Burettes, PTFE Stopcock
Supplier: VWR Collection
612-4249EA 0 INR
612-4249 612-4257 612-4258
VWR® Burettes, PTFE Stopcock
Burettes
Borosilicate glass 3,3, class AS.
With straight bore PTFE key stopcock, Schellbach stripe, with LOT certificate.
Accuracy as per AS DIN 12700 ISO 385.