VWR® Burettes, Amber Graduation
Supplier: VWR Collection
612-4227EA 551 INR
612-4227 612-4228 612-4229
VWR® Burettes, Amber Graduation
Burettes
Borosilicate glass, grade 3,3, class B.
Clear glass with glass key stopcock, amber graduation.