VWR® Burettes, Glass Key Stopcock
Supplier: VWR Collection
612-4233EA 0 INR
612-4233 612-4235 612-4237
VWR® Burettes, Glass Key Stopcock
Burettes
Borosilicate glass 3,3, class AS.
Burette with straight bore glass key stopcock, LOT certificate.
Accuracy as per class AS DIN 12700, ISO 385.