VWR® Bulb Pipettes, One Mark, Class B
Supplier: VWR Collection
612-4677EA 526 INR
612-4677 612-4678 612-4680 612-4681 612-4682 612-4683 612-4684 612-4679
VWR® Bulb Pipettes, One Mark, Class B
Pipettes Bulb Pipettes
Borosilicate glass, grade 3.3
Certifications: Accuracy as per DIN 12691, ISO 648