VWR® Bulb Pipettes, One Mark, Class A
Supplier: VWR Collection
612-4671EA 0 INR
612-4671 612-4672 612-4673 612-4674 612-4675 612-4676 612-4705 612-4707
VWR® Bulb Pipettes, One Mark, Class A
Pipettes Bulb Pipettes
Borosilicate glass, grade 3.3
  • With individual work certificate
  • Serially numbered
Certifications: Accuracy as per DIN 12691, ISO 648