VWR® Beakers, Tall Form
Supplier: VWR Collection
213-0371EA 1286 INR
213-0371 213-0372
VWR® Beakers, Tall Form
Beakers
Borosilicate glass, grade 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
With graduation and spout.
Certifications: DIN 12331, ISO 3819