Masterflex® Nylon Clamp for Sanitary Fittings, 2"; 10/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX12805-05
Your Organization's Price:
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics