Masterflex® Nylon Clamp for Sanitary Fittings, 1/2-3/4" Clamp Size; 10/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX00424-TN
Your Organization's Price:
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics