Peristaltic Pump, Masterflex®, Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
Supplier: Avantor Fluid Handling
VWR Item#MFLX07526-99
Your Organization's Price:
Adapts a variety of sensors to the MasterSense™ Sensor AUX input.
  • Designed for use with 0-20/4-20 mA or 0-5,0-10 V sensors
  • Includes 24VDC power out pins for loop powered circuits up to 50 mA
Connection cable allows for integration of variety of sensors into Masterflex® MasterSense™ pump systems. Reference MasterSense™ instruction manual for wiring diagrams and detail.