Masterflex® Fitting, Nylon, Straight, Female Luer to Low-Profile Hose Barb Adapter, 0.17" ID; 25/PK
Supplier: Avantor Fluid Handling
Ensure your process integrity with full material traceability.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance
- Connection Type: Female Luer x 0.17" ID
- Ideal for use with semi-rigid tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.