Spirax Sarco Cast Iron Float/Thermostatic Steam Trap, Parallel Config.; 3/4"NPT(F), 75 psig
Supplier: Avantor Fluid Handling
Keep steam space free of condensate to provide maximum system efficiency.
- Constructed of durable cast iron
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections. <br /><br /><B>Typical applications:</B><br />All process equipment, particularly when controlled by modulating temperature control valves<br />Unit heaters<br />Air heat coils<br />Heat exchangers<br />Steam main drip stations