Masterflex® DB15 Male Connector; for Creating Your Own Cable
Supplier: Avantor Fluid Handling
VWR Item#MFLX07595-52
Your Organization's Price:
Create your own cable for optimization of your Masterflex® drive.
  • Choose the correct connection for your Masterflex® pump
Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).