Parker Hannifin Brass Metering Valve with 1/4" NPT(M) connections, 0.125" orifice
Supplier: Avantor Fluid Handling
VWR Item#MFLX98450-33
Your Organization's Price:
Don’t worry about repeating flowrates.
  • Easily repeat flow settings with analog knob
These valves feature an analog knob for repeatable flow settings. Brass valves have Buna N O-rings; 316 SS valves have Viton® O-rings.