Parker Hannifin Brass Metering Valve with compression connections, 1/4" tube OD, 0.031" orifice
Supplier: Avantor Fluid Handling
VWR Item#MFLX98450-05
Your Organization's Price:
Don’t worry about repeating flowrates.
  • Easily repeat flow settings with analog knob
These valves feature an analog knob for repeatable flow settings. Brass valves have Buna N O-rings; 316 SS valves have Viton® O-rings.