Masterflex® Low-Flow Peristaltic Pump, Manual Control, Variable Speed, 1 to 0.03 mL/min; 12 VDC
Supplier: Avantor Fluid Handling
VWR Item#MFLX73160-00
Your Organization's Price:
Variable-flow reversible peristaltic pump.
  • Flow ranges from 0.03 to 4 ml/min
  • Run dry without damage to pump
These cost-effective peristaltic pumps provide variable flow and exceptional life performance in a compact package. Select from three different motor speeds, and three different tubing sizes to achieve flow rates from 0.03 to 4 ml/min.<br /><br />Order power supply separately. Wall mount power supply, 115 VAC/12 VDC with US standard plug sold separately as part number 73160-50. Wall mount power supply, 230 VAC/12 VDC with EU plug sold separately as part number 73160-52.
<b>Delivery information: </b>Includes silicone tube sets included; Norprene® and Viton® tubing sets available.