Masterflex® Fitting, Nylon, Straight, Female Luer to Hose Barb Adapter, 1/16" ID; 25/PK
Supplier: Avantor Fluid Handling
VWR Item#MFLX45502-00
Your Organization's Price:
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Connection Type: Female Luer x 1/16" ID
  • Ideal for use with flexible tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.